DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) Mitochondrien-Färbemittel, für die Fluoreszenzmikroskopie
Lieferant: Biotium
|
Achtung
|
70018
70018.EA
89.6
EUR
70018.
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) Mitochondrien-Färbemittel, für die Fluoreszenzmikroskopie
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide)
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) is used to stain mitochondria in live cells.
- Cellular localization: Mitochondria
- For live or fixed cells: For live/intact cells
- Assay type/options: No-wash staining, Real-time imaging
- Cell permeability: Membrane permeant
- Apoptosis/viability marker: Mitochondrial potential
- Colors: Red
- Excitation/Emission: 461/589 nm
- Store at 4°C and protect from light, especially in solution
Formel:
C₁₇H₂₁IN₂ Molecular Weight: 380,273 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00011994 CAS-Nummer: 3785-01-1 EINECS: 223-248-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...