Calcium Ionophore IV - Cocktail for membranes, Selectophore™ Material für Ionensensoren, Supelco®

Lieferant: Merck
Gefahr

70356-1ML-F
70356-1ML-FEA 215 EUR
70356-1ML-F
Calcium Ionophore IV - Cocktail for membranes, Selectophore™ Material für Ionensensoren, Supelco®
Calcium Ionophore IV - Cocktail for membranes

Composition:
Calcium ionophore IV, 0.072 wt. % (21198)
2-Nitrophenyl octyl ether, 4.748 wt. % (73732)
Poly(vinyl chloride), 2.379 wt. % (81392)
Potassium tetrakis(4-chlorophenyl)borate, 0.022 wt. % (60591)
Tetrahydrofuran, 92.780 wt. % (87369)


Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Flammpunkt: –18 °C
UN: 2056
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

APPEARANCE (COLOR) Colorless to Yellow and Colorless to Green-Yellow
APPEARANCE (FORM) Liquid
APPLICATION TEST CORRESPONDS TO REQUIREMENTS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR