Ethylenglycol, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Glycol, 1,2-Dihydroxyethan, 1,2-Ethandiol
7037.1000EA
71.98
EUR
7037.1000
7037.2500
7037.9025
Ethylenglycol, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Ethylenglycol
Formel:
HOCH₂CH₂OH Siedepunkt: 196…198 °C (1013 hPa) Schmelzpunkt: –13 °C Dichte: 1,115 g/cm³ (20 °C) Flammpunkt: 116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00002885 CAS-Nummer: 107-21-1 EINECS: 203-473-3 REACH: 01-2119456816-28 Merck Index: 13,03832 |
|
Spezifikation Testergebnisse
Assay (by GC) | min. 99.0 % |
Acidity (as CH3COOH) | max. 0.01 % |
Boiling Range | 194.0 - 200.0 °C |
Boiling Point | 195 - 198 °C |
Color (APHA) | max. 10 |
Density (g/ml) at 20°C | 1.110 - 1.114 |
Residue after Ignition | max. 0.005 % |
Water (H2O) | max. 0.2 % |
Trace Impurities (in ppm) | |
Chloride (Cl) | max. 5 |
Iron (Fe) | max. 0.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...