Glycerin, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: 1,2,3-Propantriol, Glycerol

7044.2500EA 255.26 EUR
7044.2500 7044.1000 7044.9025
Glycerin, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Glycerin
Formel: HOCH₂CH(OH)CH₂OH
Siedepunkt: 290 °C (1013 hPa)
Schmelzpunkt: 18,6 °C
Dichte: 1,26 g/cm³ (20 °C)
Flammpunkt: 177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004722
CAS-Nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay min. 99.5 %
Silver Reducing Substances passes test
Color passes test
Density (g/ml) at 25°C min. 1.255
Fatty Acid Esters max. 0.05 %
Neutrality passes test
Residue after Ignition max. 0.005 %
Substances Darkened by H2SO4 passes test
Sulfate (SO4) max. 0.001 %
Trace Impurities (in ppm)
Chloride (Cl) max. 5
Heavy Metals (as Pb) max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR