Compact system for 100 ml extraction, KEX 100/KEX 100 F, behrotest®
Lieferant: BEHR
B00217710 B00217708
705-0002EA 962 EUR
705-0002 705-0001
Compact system for 100 ml extraction, KEX 100/KEX 100 F, behrotest®
Extraktoren Extraktionsapparate, nach Soxhlet
Complete compact extraction system with basic rack, heating device, holder, tubing and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
  • Practical holders for condenser and extraction adapter for secure placement between extractions
  • Extractor sizes from 30 to 500 ml
  • Condenser with screw connections
  • Practical stand for holding condensers securely between extractions
  • Holders for secure fixation of extraction adapters after removal of the sample vessels
  • S models: The use of extractors with a stopcock does away with the need for any additional distillation after extraction
Infinitely variable heating regulation. After the extraction cycle, the compact system removes the solvent, by means of a discharge valve on the extractor directly into the reservoir. behr equipment for Soxhlet extraction complies with the various requirements of daily laboratory routines.