Bromphenolblau
Lieferant: Hach
Synonyme:
3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid
1455099
705-0070EA
40.7
EUR
705-0070
Bromphenolblau
Bromphenolblau
Formel:
C₁₉H₁₀Br₄O₅S Schmelzpunkt: 273 °C Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005875 CAS-Nummer: 115-39-9 EINECS: 204-086-2 Merck Index: 13,01429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...