Test strips
Lieferant: Hach
2745250
2744850
2745650
2744940
2751340
HACH2745250EA
35.6
EUR
HACH2745250
705-0185
705-0189
705-0186
705-0190
Test strips
Umwelt-Prüfkits
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
- Portable - ideal for field testing of surface water
- Easy to use