Test strips
Lieferant: Hach
2745250 2744850 2745650 2744940 2751340
HACH2745250EA 35.6 EUR
HACH2745250 705-0185 705-0189 705-0186 705-0190
Test strips
Umwelt-Prüfkits
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use