Test strips, chloride, QUANTOFIX®
Lieferant: MACHEREY-NAGEL
91321
705-0321EA 40 EUR
705-0321
Test strips, chloride, QUANTOFIX®
Umwelt-Prüfkits
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of chloride in solutions. The easy dip-and-read-procedure provides a reliable result within 1 minute.
Chloride ions occur in all natural waters. Their concentration depends on the geological and local situation. In waste waters and polluted rivers the chloride concentration can reach high values. In combination with sodium, chloride forms NaCl or table salt.
Gradation: 0 · 500 · 1000 · 1500 · 2000 · ≥3000 mg/L Cl-
Colour: brown → yellow