Test strips, molybdenum, QUANTOFIX®
Lieferant: MACHEREY-NAGEL
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage color scale
- Always ready to use: No accessories necessary for evaluation
- Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of molybdenum in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within 1 minute one gets a reliable result.
Molybdenum tests are mainly used for coolant water and boiler feed water. Here, Molybdenum salts act as corrosion inhibitors, either directly or as part of an anticorrosive. The careful control of the Molybdenum level is important to ensure optimal corrosion prevention. Also, the level of the anticorrosive used can be easily controlled.
Gradation: 0 · 5 · 20 · 50 · 100 · 250 mg/L Mo⁶+
Colour: white → green