Infrared rapid digestion devices (programmable) for nitrogen determination according to Kjeldahl, behrotest® InKjel
Lieferant: BEHR
B00218108 B00218105 B00218106 B00218107 B00497540
705-0437EA 0 EUR
705-0437 705-0434 705-0435 705-0436 BEHRB00497540
Infrared rapid digestion devices (programmable) for nitrogen determination according to Kjeldahl, behrotest® InKjel
Aufschlusssysteme Aufschluss-Systeme
behrotest® InKjel digestion models are equipped with a high performance infrared heater.
  • Direct heating of the samples by the infrared heater prevents the endlessly long heating times required using conventional heating block systems
  • High quality quartz heaters instead of the common steel tube heater ensures particularly homogeneous heating in all sample positions
  • behr one-button operation for extremely easy and quick programming
  • Menu navigation in language of the country
Quality and position of the behr infrared heater guarantee the user identical heating phases and digestion temperatures in all sample positions. This also applies to the double-row arrangement for racks of 12 samples. The glass digestion vessels are suspended in the rack and are not deposited at the bottom of the digestion unit. Therefore, the digestion units are less susceptible to breakage than in an aluminium heating block.
This makes the behrotest® InKjel the ideal digestion system for nitrogen determination according to Kjeldahl and other high temperature digestions. In Kjeldahl digestions, the temperature is adjusted through the boiling point of sulphuric acid.