Determination of crude cellulose fibre, ADF and NDF, FibreBag
Lieferant: GERHARDT
10-0015 10-0013
705-0469EA 6900 EUR
705-0469 705-0467
Determination of crude cellulose fibre, ADF and NDF, FibreBag
Extraktoren Rohfaserbestimmungssysteme
For rapid determination in accordance with Weender, Van Soeste et al. Use fine mesh bags to weigh the sample. A glass diffuser is then placed inside the bag, allowing the reagents to diffuse freely. Six samples can be processed simultaneously: clean, boil in sulphuric acid, rinse, boil in KOH, rinse again and dry.
  • Considerably reduces reagent consumption
  • Complies with official methods
A six-piece pack includes the following equipment: heating plate, six-piece carousel, handle for the carousel, six glass diffusers, plate stand, glass condenser and tubing.
Zubehör auf Anfrage