Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ4000
Lieferant: Thermo Orion
AQ4000
705-0731EA 1410 EUR
705-0731
Portable multiparameter colorimeter, Orion™ AQUAfast™ AQ4000
Kolorimeter Mehrparameter-Kolorimeter
Measure common colorimetric water and wastewater parameters using the Orion™ AQUAfast™ AQ4000 colorimeter with 65 preprogrammed methods, automatic selection of four wavelengths and timer.
  • Identifies the species to be measured
  • Ensures error-free readings
  • Selects the programme and initiates the measurement
  • Guides user through the setup and measurement with simple operation
  • User can customise calibration and create completely custom methods
  • Up to 12 data points can be used to define custom method
  • Store a maximum of 10 methods in the colorimeter memory
Auto-Test ampoules with Auto-ID barcodes can be used to simplify sample preparation and tests. Ampoules contain pre-measured liquid reagent and fill with the exact volume of sample, colorimeter reads the barcode and automatically identifies the parameter and initiates the test.