Comparator kits, multiparameter for water testing, Comparator 2000+
Lieferant: Lovibond
413570 413580
705-0787EA 2075.2 EUR
705-0787 705-0362
Comparator kits, multiparameter for water testing, Comparator 2000+
Kolorimeter Komparator-Sets
Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.
  • Compact and portable
  • Simple to use yet uncompromising in terms of precision and reproducibility of results
Lieferumfang: Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond® tablet reagents (for 100 measurements). The AF368 is not supplied with reagents.