UV/Vis-Spektralphotometer, DR6000
Lieferant: Hach
LPV4419900011 LPV4419900002 LPV4419900001
705-0913EA 13420 EUR
705-0913 705-1161 705-1160
UV/Vis-Spektralphotometer, DR6000
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
UV/Visible spectrophotometers with radio frequency (RFID) technology to ensure high speed wavelength scanning across the UV and visible spectrum.
  • Improved laboratory performance thanks to 240 available methods
  • Transmittance (%), absorbance and concentration, scanning
  • RFID technology allows sample traceability
  • Optimal data management and reliable results
  • Key methods are possible including COD, chlorine, nitrates and ammonia
  • High quality reagents formulated to ensure outstanding performance and results
  • Software Quality Assurance (SQA) for scheduling, documenting and interpreting
Some applications that are suitable for enzymology and colorimetry, open up further application opportunities including drinking water and brewery analysis.
For a list of popular test kits for the DR6000 instrument, please see the COMPLEMENTARY PRODUCTS tab. For a more comprehensive selection of available test kits, please search the website using the search term ETK.
Bestellinformation: Some application packages are suitable for enzymology and colorimetry, opening up further applications including drinking water and brewing.
Lieferumfang: Supplied with cell adapter, dust cover, power cord and user manual.