Test strips, quaternary ammonium compounds (QUAT), QUANTOFIX®
Lieferant: MACHEREY-NAGEL
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale
- Always ready to use: No accessories necessary for evaluation
- Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of quaternary ammonium compounds (QUAT) in solutions. The easy dip-and-read-procedure provides a reliable result within 15 seconds.
QUAT based disinfectants are frequently used for the disinfection of medical devices, surfaces and closed cooling cycles. These test strips allow convenient monitoring to decide, if the concentration of the disinfectant is sufficient. This ensures an optimal disinfection. They are calibrated for Benzalkonium chloride. The instructions contain conversion factors for many other quaternary ammonium compounds. These test strips are the perfect tool to monitor the QUAT concentration in a solution and provide safe results quickly and easily.
Gradation: 0 · 10 · 25 · 50 · 100 · 250 · 500 · 1000 mg/l benzalkonium chloride
Colour: yellow → blue-green