Chromium standard solutions
Lieferant: Hach
1425610 81042H 1466442
705-2007EA 79.3 EUR
705-2007 HACH81042H HACH1466442
Chromium standard solutions
Standards Kationischer Standards Chrom-Standards
Standard solution for accuracy checks in total chromium determinations.
  • Traceable to NIST