Standard solution, detergent
Lieferant: Hach
1427110
705-2016EA 59.4 EUR
705-2016
Standard solution, detergent
Standards Umweltanalyse-Standards
For accuracy checks in detergents (surfactants) determinations.