Wasser zur Verdünnung Organik-frei

Lieferant: Hach

2641549
705-2037EA 83.1 EUR
705-2037
Wasser zur Verdünnung Organik-frei
Wasser

Used for UV-254 and TOC methods.

Formel: H₂O
MDL: MFCD00011332
CAS-Nummer: 7732-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR