2-Propanol Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

7065-33EA 0 EUR
7065-33 7065-38 7065-39
2-Propanol Ph. Eur., Macron Fine Chemicals™
2-Propanol
Formel: (CH₃)₂CHOH
MDL: MFCD00011674
CAS-Nummer: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR