Silbernitrat 4,791 g/L 0.0282 mol/l (0.0282 N) in wässriger Lösung, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
Synonyme:
Höllenstein
7095.1000EA
0
EUR
7095.1000
Silbernitrat 4,791 g/L 0.0282 mol/l (0.0282 N) in wässriger Lösung, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Silbernitrat
1 ml = 1 mg Cl
Formel:
AgNO₃ Molecular Weight: 169,87 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003414 CAS-Nummer: 7761-88-8 EINECS: 231-853-9 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | |
Particle Size Distribution: APD, microns | 47 - 61 |
Specific Surface Area, m/g, For Information Only | |
Result based on the bulk bonded phase - Carbon (C), For Information Only | |
Surface Coverage - Based on %C, For Information Only | |
Result based on the solid phase extraction columns- Extraction Residue | ≤ 0.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...