Transystem™ - soft aluminium applicator, rayon wrapped minitip, amies with charcoal, choked tube, 139 mm, 50×10, individually wrapped, Copan Transystem®, Verschluss-Farbe: Grün
Lieferant: Copan
Transystem™ - soft aluminium applicator, rayon wrapped minitip, amies with charcoal, choked tube, 139 mm, 50×10, individually wrapped, Copan Transystem®, Verschluss-Farbe: Grün