Abklatschplatten, gammabestrahlt, dreifach verpackt, ICR (Isolator, Reinraum)
Lieferant: MERCK LIFE SCIENCE
1462310200
1465520020
1461950200
1465520200
1461950020
1462310020
1465010200
1465380020
1465380200
1465390200
710-1893EA
1120
EUR
710-1893
HEIP1465520020
HEIP1461950200
HEIP1465520200
710-1891
710-1892
HEIP1465010200
HEIP1465380020
HEIP1465380200
HEIP1465390200
Abklatschplatten, gammabestrahlt, dreifach verpackt, ICR (Isolator, Reinraum)
Medien für die Mikrobiologie
Abklatschplatten für die Mikrobiologie
ICR/ICRplus contact plates are triple-bagged and gamma irradiated to ensure the highest level of safety when monitoring surfaces in isolators and critical clean rooms for microorganisms.
- Data matrix barcode on each plate for paperless and secure identification of individual plates
- CLOSED and VENT closure options of ICRplus contact plates for safe transportation and variation of incubation conditions
- Convenient room temperature stability allows storage at site of use