Readyplate XLD Agar acc ISO 6579
Lieferant: MERCK LIFE SCIENCE
1467510020
710-2268EA
124
EUR
710-2268
Readyplate XLD Agar acc ISO 6579
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
Application: Identification of Salmonella
- Packaging: 90 mm plate (20 plates per box)
- Synonym: Xylose Lysine Deoxycholate Agar
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...