Mini magnifier
Lieferant: HEATHROW
HS15092D
710-4001EA 216 EUR
710-4001
Mini magnifier
Lupen
This mini magnifier can be used as an accessory for the eCount® or can be stand-alone.
Mini magnifier enlarges objects for easier viewing. Swivelling, moulded lens, with 1,75× magnification. Lens measures 185×100 mm; stand measures 207 mm (height).
Bestellinformation: Mini LED light box must be purchased separately.