Zinn(II)iodid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 10294-70-9
Molecular Formula: SnI2
Formula Weight: 372.50
Physical Form: -6 Mesh Powder
Density: 5.28
Appearance: Yellow to red
Melting Point: 320°
Boiling Point: 720° dec.
Solubility: Soluble in benzene, CS2, chloroform, and solutions of alkali chlorides or iodides. Decomposes in water
Merck Reference: 14,8785
UN No.: 3260
MDL No.: MFCD00049545
Formel:
I₂Sn Siedepunkt: 720 °C (Zersetzung) Schmelzpunkt: 320 °C Dichte: 5,28 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049545 CAS-Nummer: 10294-70-9 EINECS: 233-667-3 UN: 3260 ADR: 8,III Merck Index: 14,8785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...