Kaliumhydrogensulfat, powder_fused, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Kaliumbisulfat

7112-20EA 0 EUR
7112-20
Kaliumhydrogensulfat, powder_fused, AR® ACS, Macron Fine Chemicals™
Kaliumhydrogensulfat
Formel: KHSO₄
Molecular Weight: 136.17 g/mol
Schmelzpunkt: 214
Dichte: 2.32 g/cm³ (20 °C)
MDL: MFCD00011404
CAS-Nummer: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 37.5 - 38.6 %
ACS - Acidity (as H₂SO₄) 37.5 - 38.6 %
ACS - Water (H₂O) ≤2.5 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %
ACS - Sodium (Na) ≤0.01 %
ACS - Insoluble Matter ≤0.01 %
ACS - Ammonium Hydroxide Precipitate ≤0.01 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Magnesium (Mg) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR