(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥99,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®

Lieferant: Merck
Achtung

Synonyme: 1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide, N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (2S)-

M-Clarity™ Qualitätsstufe = MQ100

71478-50MG
71478-50MGEA 218 EUR
71478-50MG
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥99,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide

Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide (FDAA) is a chiral derivatizing agent (CDA), has high enantioselectivity but low sensitivity as compared to other CDAs. It is generally used to assign the stereochemistry of amino acids in trace amounts.

Formel: C₉H₉FN₄O₅
Molecular Weight: 272,192 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00042049
CAS-Nummer: 95713-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fed605b8a78203691fd16fb7f61c0eec4e7b4c3&vid=7377acc913042e6cee274a4caf6b9e118cae57a3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR