Natriumdihydrogenphosphat, wasserfrei ≥99,999% (Metall-Basis), TraceSELECT™ für die Spurenanalyse, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Natriumphosphat monobasisch

71492-100G
71492-100GEA 352.1 EUR
71492-100G
Natriumdihydrogenphosphat, wasserfrei ≥99,999% (Metall-Basis), TraceSELECT™ für die Spurenanalyse, Fluka™
Natriumdihydrogenphosphat

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formel: NaH₂PO₄
Molecular Weight: 119,98 g/mol
Schmelzpunkt: 650 °C
Dichte: 2,36 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00003527
CAS-Nummer: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Bestellen

Spezifikation Testergebnisse

Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Cesium (Cs) Max. 5 ppm
Copper (Cu) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Mercury (Hg) Max. 5 ppb
Iridium (Ir) Max. 2 ppm
Potassium (K) Max. 5 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 1 ppm
Magnesium (Mg) Max. 0.5 ppm
Manganese (Mn) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rubidium (Rb) Max. 5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.2 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.5 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.1 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.01 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm
Total nitrogen (as N) Max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR