Natriumhydrogensulfat Monohydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a.

Lieferant: Honeywell Chemicals
Gefahr

71657-500G
71657-500GEA 61.19 EUR
71657-500G
Natriumhydrogensulfat Monohydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a.
Natriumhydrogensulfat Monohydrat
Formel: NaHSO₄·H₂O
Siedepunkt: Decomposes on heating
Schmelzpunkt: 58 °C
Dichte: 2,12 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149210
CAS-Nummer: 10034-88-5
EINECS: 231-665-7
ADR: 8,III
Merck Index: 13,08658

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 20 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 5 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR