Natriumhydroxid 97,0 Titrimetric, pellets, Purum p.a. analytical reagent
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Caustic soda
71692-5KG
71692-5KGEA
171.1
EUR
71692-5KG
Natriumhydroxid 97,0 Titrimetric, pellets, Purum p.a. analytical reagent
Sodium hydroxide
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 1.390 °C (1.013 hPa) Schmelzpunkt: 319 °C Dichte: 2.13 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8 II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Spezifikation Testergebnisse
Vapor Pressure | 3 mmHg ( 37 °C) |
Assay | Min. 97 % |
Assay of Na₂CO₃ | Max. 0.8 % |
Silver (Ag) | Max. 0.001 % |
Aluminium (Al) | Max. 0.001 % |
Calcium (Ca) | Max. 0.005 % |
Cadmium (Cd) | Max. 0.005 % |
Cobalt (Co) | Max. 0.005 % |
Copper (Cu) | Max. 0.005 % |
Iron (Fe) | Max. 0.001 % |
Mercury (Hg) | Max. 0.00001 % |
Potassium (K) | Max. 0.1 % |
Nickel (Ni) | Max. 0.001 % |
Lead (Pb) | Max. 0.001 % |
Zinc (Zn) | Max. 0.005 % |
Chloride (Cl) | Max. 0.005 % |
Total N | Max. 0.0005 % |
Phosphate (PO₄) | Max. 0.001 % |
Silicate (as SiO₂) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...