Natriumhydroxid, tested according to Ph.Eur., Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Caustic soda
71694-5KG
71694-5KGEA
139.74
EUR
71694-5KG
Natriumhydroxid, tested according to Ph.Eur., Fluka™
Sodium hydroxide
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 1.390 °C (1.013 hPa) Schmelzpunkt: 319 °C Dichte: 2.13 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8 II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Spezifikation Testergebnisse
Assay | 97.0 - 100.5 % |
pH | Min. 11.0 |
Appearance of the solution | complying |
Iron (Fe) | Max. 10 ppm |
Heavy metals (as Pb) | Max. 20 ppm |
Assay of Na₂CO₃ | Max. 2 % |
Chloride (Cl) | Max. 100 ppm |
Sulfate (SO₄) | Max. 100 ppm |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...