Benzethoniumchlorid 0.004 mol/l, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: Hyamine® 1622

7170.1000EA 37.05 EUR
7170.1000 7170.2500
Benzethoniumchlorid 0.004 mol/l, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Benzethoniumchlorid
Formel: C₂₇H₄₂ClNO₂
MDL: MFCD00011742
CAS-Nummer: 121-54-0
EINECS: 204-479-9

Bestellen

Spezifikation Testergebnisse

Titer 0.0039 - 0.0041 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR