Natriumiodat ≥99,5% (RT), Puriss. p.a., Fluka™

Lieferant: Honeywell Chemicals
Gefahr

71702-25G
71702-25GEA 69.95 EUR
71702-25G
Natriumiodat ≥99,5% (RT), Puriss. p.a., Fluka™
Natriumiodat
Formel: INaO₃
Molecular Weight: 197,89 g/mol
Schmelzpunkt: 420…430 °C
Dichte: 4,28 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003533
CAS-Nummer: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 99.5 - 101.0 %
pH (5 %, 20°C) 5.0 - 8.0
Solution 10 g + 200 mL (H₂O) complying
Calcium (Ca) Max. 100 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 50 ppm
Iodide (I) Max. 20 ppm
Total N Max. 50 ppm
Chlorate, Chloride, Bromide, Bromate (as Cl) Max. 200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR