Kieselgel, Supelco®
Lieferant: Merck
717185-1KG
717185-25KG
717185-5KG
717185-1KGEA
110
EUR
717185-1KG
717185-25KG
717185-5KG
Kieselgel, Supelco®
Silica gel
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 Schmelzpunkt: 1710 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 63231-67-4 REACH: 01-2119379499-16 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...