Sodium ionophore III ≥98,0% (durch HPLC), Selectophore™, Funktion getestet Ionophor, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide, ETH 2120
M-Clarity™ Qualitätsstufe = MQ100
Sodium ionophore III is generally used for preparing ion-selective electrode.
It may be used in the preparation of vulcanizing-type silicone rubber-based sodium-selective membrane electrodes.
Ionophore suitable for the assay of Na+-activity in blood, plasma, serum. etc. with a solvent polymeric membrane electrode.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₃₄H₅₂N₂O₄ |
MDL:
MFCD00077941 CAS-Nummer: 81686-22-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...