Sodium ionophore III ≥98,0% (durch HPLC), Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck
Achtung

Synonyme: N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide, ETH 2120

M-Clarity™ Qualitätsstufe = MQ100

71734-250MG 71734-50MG
71734-250MGEA 1290 EUR
71734-250MG 71734-50MG
Sodium ionophore III ≥98,0% (durch HPLC), Selectophore™, Funktion getestet Ionophor, Supelco®
Sodium ionophore III

Sodium ionophore III is generally used for preparing ion-selective electrode.


It may be used in the preparation of vulcanizing-type silicone rubber-based sodium-selective membrane electrodes.


Ionophore suitable for the assay of Na+-activity in blood, plasma, serum. etc. with a solvent polymeric membrane electrode.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₄H₅₂N₂O₄
MDL: MFCD00077941
CAS-Nummer: 81686-22-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c76d445272a547e3546b70efd14f4ccd82b81f0a&vid=2299602cf35092c346a4efc90b7124da99d7ac7a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR