Sodium ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
Sodium ionophore for assaying Na+-activities with grafted wire sodium-selective electrodes. It can be used either in a liquid membrane or in a PVC solid membrane
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₃₄H₆₂O₁₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD00010527 CAS-Nummer: 80403-59-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...