Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate, Selectophore™, Funktion getestet Ionophor, Supelco®
Lieferant: Merck
Synonyme:
4-tert-Butylcalix[4]arene-O,O',O'',O'''-tetraacetic Acid Tetraethyl Ester, Tetraethyl 2,2',2'',2'''-((15,35,55,75-tetra-tert-butyl-1,3,5,7(1,3)-tetrabenzenacyclooctaphane-12,32,52,72-tetrayl)tetrakis(oxy))tetraacetate
M-Clarity™ Qualitätsstufe = MQ100
Sodium ionophore X is generally used in the preparation of ion-selective electrode (ISE).
It may be used in a comparison study of ISE-based electronic tongue for direct and two-stage data analysis on Artificial Neural Network (ANN), Partial Least Squares-Discriminant Analysis (PLS-DA)) and Principal Components Analysis (PCA).
Ionophore employed in solid-state and polymeric membrane sodium-selective electrodes.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₆₀H₈₀O₁₂ Molecular Weight: 993.29 g/mol |
MDL:
MFCD00145373 CAS-Nummer: 97600-39-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...