Natriumperoxid ≥95%, kleine Perlen ACS für Wurzschmitt-Methode, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
di-Natriumperoxid
71883-1KG
71883-1KGEA
751.78
EUR
71883-1KG
Natriumperoxid ≥95%, kleine Perlen ACS für Wurzschmitt-Methode, Fluka™
Natriumperoxid
Formel:
Na₂O₂ Molecular Weight: 77,98 g/mol Siedepunkt: Decomposes on heating Schmelzpunkt: 660 °C Dichte: 2,8 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003497 CAS-Nummer: 1313-60-6 EINECS: 215-209-4 UN: 1504 ADR: 5.1,I Merck Index: 13,08728 |
Spezifikation Testergebnisse
Assay | 95.0 - 105.0 % |
Silver (Ag) | Max. 5 ppm |
Aluminium (Al) | Max. 20 ppm |
Barium (Ba) | Max. 5 ppm |
Bismuth (Bi) | Max. 5 ppm |
Calcium (Ca) | Max. 500 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 200 ppm |
Lithium (Li) | Max. 5 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 5 ppm |
Molybdenum (Mo) | Max. 5 ppm |
Nickel (Ni) | Max. 10 ppm |
Lead (Pb) | Max. 5 ppm |
Strontium (Sr) | Max. 10 ppm |
Thallium (Tl) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Heavy metals (as Pb) | Max. 0.002 % |
Chloride (Cl) | Max. 20 ppm |
Phosphate (PO₄) | Max. 5 ppm |
Sulfate (SO₄) | Max. 10 ppm |
Total N | Max. 30 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...