Staining Racks
Lieferant: DIXON SCIENTIFIC
406019000
720-0096EA 499 EUR
720-0096
Staining Racks
Gestelle Färbegestelle
Comprises two brass end-plates fitted with levelling screws and clips.
  • For holding two glass rods with diameter of ∼8 to 9 mm
Bestellinformation: Rods are not included.