Cervical brushes, Cytotak™ Transwab®, labelled
Lieferant: MEDICAL WIRE EQUIPMENT
MW148
720-0136EA 346 EUR
720-0136
Cervical brushes, Cytotak™ Transwab®, labelled
Bürsten
Nylon-bristle cytology sampling brushes for cervical smears.
  • Sterile, packed in labelled tube
  • Plastic shaft is non toxic and safe to use
  • Tapered spiral configuration assists access to narrow opening
  • Gentle rotation creates a brushing action against skin surface for enhanced recovery of cellular material
Shaft/head: Plastic/nylon
Verpackung: Individually packed.