Erythrosin 239 Färbemittel für Histologie

Lieferant: RAL
Achtung

Synonyme: C.I. 45430, Erythrosin 239

312820-0100
720-0179EA 0 EUR
720-0179
Erythrosin 239 Färbemittel für Histologie
Erythrosin B Dinatriumsalz

The Erythrosine 239 is used in the staining by the Haematoxylin-Erythrosine bichromic for routine staining of histological sections. The Erythrosin stains collagen very pale pink, cytoplasms pink to red and the leaf springs and the red cells in bright pink.

Formel: C₂₀H₆I₄Na₂O₅
Molecular Weight: 879,86 g/mol
MDL: MFCD00144257
CAS-Nummer: 16423-68-0
EINECS: 240-474-8
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR