Carbolic auramine
Lieferant: RAL
361430-1000
720-0345EA 69.3 EUR
720-0345
Carbolic auramine
Carbolic auramine
RAL Auramine staining kit allows fluorescence detection of Acid Fast Bacilli (AFB). The A.F.B appear fluorescent-yellow on the blue background preparation.
Used in Auramine staining kit Ref 359500
IVDLogo