Myeloperoxydase kit for haematology
Lieferant: RAL
361610-0000
720-0352EA
99.24
EUR
720-0352
Myeloperoxydase kit for haematology
Myeloperoxydase kit for haematology
Staining Kit for the Differentiation of Leukaemia.
Peroxidase granulations form an insoluble bright red compound with pyronin in presence of Alphanaphtol and Hydrogen Peroxide. The nuclei of leukocytes stain blue with Mayer Haematoxylin. This kit is benzidine-free. Benzidine is replaced by Alphanaphtol-Pyronin.