Staining jars acc. to Coplin, WHEATON®
Lieferant: DWK Life Sciences
276840
720-0700EA 109 EUR
720-0700
Staining jars acc. to Coplin, WHEATON®
Dosen Färbeküvetten
Staining jars, made of soda-lime glass, with extra tall lid.
It holds five single 75×25 mm and 75×100 mm slides vertically, or 10 slides back to back.