Uranin, rein für Hydrologie

Lieferant: RAL

Synonyme: 6-Hydroxy-9-(2-carboxyphenyl)-(3H)-xanthen-3-on Dinatriumsalz, 6-Hydroxy-9-(2-carboxyphenyl)- (3H)-xanthen-3-on Dinatriumsalz, Fluoreszein Dinatriumsalz, Fluorescein Natriumsalz, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)-benzoesäure Dinatriumsalz, Dinatrium-2-(3-oxo-6-oxidoxanthen-9-yl)benzoat, 6-Hydroxy-9-(2-carboxyphenyl)-(3H)-3-xanthenon Dinatriumsalz, Fluorescein-Natrium, Resorcinphthalein Dinatriumsalz, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoesäure Dinatriumsalz, Fluorescein Dinatriumsalz, 3′,6′-Dihydroxyspiro[2-benzofuran-3,9′-xanthen]-1-on Dinatriumsalz, 9-(2-Carboxyphenyl)-6-hydroxy-(3H)-3-xanthenon Dinatriumsalz

313150-1000
720-1067EA 250.04 EUR
720-1067
Uranin, rein für Hydrologie
Uranin

Hydrological researches, tracer in aqueous medium, various industrial uses. A good visual detection is obtained for concentrations around 5 to 10 mg/L.

Formel: C₂₀H₁₀Na₂O₅
Molecular Weight: 376.28 g/mol
MDL: MFCD00167039
CAS-Nummer: 518-47-8
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR