CytoRAL fixative for cytological smears, RAL
Lieferant: RAL
361400-0150
720-1153EA
8.92
EUR
720-1153
CytoRAL fixative for cytological smears, RAL
Färbemittel
Mikroskopie- und Färbereagenzien
CytoRAL is used for cervical smears, urine, sputa and broncho-alveolar fluids.
It lays a protective film on the smears, keeping a good hydration before staining and without any alteration of the original cells aspect. With a large dispersion ratio the spray covers the entire slide area.