Crystal violet oxalate acc. Gram-Hucker
Lieferant: RAL
361490-2500
720-1161EA 52.5 EUR
720-1161
Crystal violet oxalate acc. Gram-Hucker
Crystal violet oxalate
Hücker ammonium oxalate crystal violet
Used in Gram-Hücker R Kit (Ref 361520)
IVDLogo