Safranin O acc. Gram-Hücker
Lieferant: RAL
Synonyme:
3,7-Diamino-2,8-dimethyl-5-phenylphenaziniumchlorid, Basisrot 2
Used in Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria.
Used in Gram-Hücker R Kit Ref 361520
Gram-Hücker staining is based on the same principle than the Gram-Nicolle staining. The structure of the bacterial walls and their permeability to alcohol help differentiate the Gram-negative bacteria from the Gram-positive ones. In this technique, Carbolic Gentian Violet is replaced by Crystal Violet Oxalate (that does not require filtration) and Ziehl-Carbolic Fuchsin by Safranin.
Formel:
C₂₀H₁₉ClN₄ Molecular Weight: 350,85 g/mol |
MDL:
MFCD00011759 CAS-Nummer: 477-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...