Copro-Duo Kit (2×24) The complete kit for a clean and reliable coprology
Lieferant: RAL
Kit Copro-Duo contains all the necessary consumables for both MIF and Bailenger concentration methods. It makes it possible to concentrate all the existing parasitic elements in a small volume.
Kit Copro-Duo is complete and ready to use. It contains all the necessary reagents and disposable utensils.
Components - Name (ref) Packing
MERCUROTHIOLATE FORMALIN (M.F SOLUTION) (Ref 365020) 400 ml
LUGOL CONCENTRATED SOLUTION (Ref 367210) 6 ml
ACETO-ACETIC BUFFER PH=5.0 ACCORDING TO BAILENGER (Ref 366220) 400 ml
48 SPATULAS
48 SEDIMENTATION TUBES (30 ml)
48 CENTRIFUGATION TUBES (10 ml)
24 TUBE RACKS
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...